Order Entry
Italy
ContactUsLinkComponent
Silicon tetraacetate, anhydrous 99%
Corrosive
Silicon tetraacetate, anhydrous 99%
Catalog # APOSIN3184-50G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Silicon tetraacetate, anhydrous 99%
Catalog # APOSIN3184-50G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :264.261 g/mol
Melting Pt:111…115 °C
Storage Temperature:Freezer
UN:3261
ADR:8,II
 :GHS05
CAS Number:562-90-3
Formula:C₈H₁₂O₈Si
MDL Number:MFCD00026184