Selenium dioxide 99,9%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:7446-08-4
- Formula:SeO₂
- MDL Number:MFCD00003562
- Molecular Weight:110.96 g/mol
- Melting Pt:340…350 °C (subl.)
- Density:3.95 g/cm³ (25 °C)
- Storage Temperature:Ambient
- UN:3283
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:7446-08-4
- Formula:SeO₂
- MDL Number:MFCD00003562
- Molecular Weight:110.96 g/mol
- Melting Pt:340…350 °C (subl.)
- Density:3.95 g/cm³ (25 °C)
- Storage Temperature:Ambient
- UN:3283
- ADR:6.1,III



