Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Palladium
Palladium on carbon 10% Pd, powder
Palladium on carbon 10% Pd, powder
APOSIN2843-25G
:
Apollo Scientific
:
Palladium on carbon 10% Pd, powder
APOSIN2843-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
106.420 g/mol
:
Ambient
:
1325
:
4.1,III
:
GHS02
:
GHS07
:
7440-05-3
:
Pd
:
MFCD00011167