Phosphorus(III) bromide 99%
Catalog # APOSIN2839-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Phosphorus tribromide
:
:270.69 g/mol
Melting Pt:–40 °C
EINECS:232-178-2
UN:1808
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7789-60-8
Formula:PBr₃
MDL Number:MFCD00011436
Boiling Pt:175 °C
Density:2.852 g/cm³ (25 °C)
ADR:8 II
Merck Index:12,07514