



Nickel(II) chloride hexahydrate 99%
Catalog # APOSIN2680-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:237.69 g/mol
Storage Temperature:Ambient
EINECS:616-576-7
UN:3288
ADR:6.1,III
:GHS06
:GHS07
:GHS08
:GHS09
CAS Number:7791-20-0
Formula:NiCl₂·6H₂O
MDL Number:MFCD00149809