

Gallium trinitrate hydrate 99.999%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69365-72-6
:
Formula:GaN₃O₉·H₂O
MDL Number:MFCD00149723
:273.75 g/mol
Melting Pt:110 °C (dec.)
Storage Temperature:Ambient
EINECS:236-815-5
UN:1477
ADR:5.1,II
:GHS03
:GHS07
Frequently Bought Together