
Barium acetate, Reagent Grade
Catalog # APOSIN1174-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:255.42 g/mol
Melting Pt:725 °C
Density:2.47 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:208-849-0
UN:1564
ADR:6.1,III
:GHS07
CAS Number:543-80-6
Formula:(H₃CCOO)₂Ba
MDL Number:MFCD00012447