Order Entry
Italy
ContactUsLinkComponent
GW 4869 dihydrocloride
GW 4869 dihydrocloride
Catalog # APOSBISN0156-10MG
Supplier:  Apollo Scientific
CAS Number:  
GW 4869 dihydrocloride
Catalog # APOSBISN0156-10MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 mg
  • Seq:
    2275
  • Environmentally Preferable:

Specifications

About this item

Synonyms: GW4869, p-Benzenediacrylanilide, 4′,4′′-di-2-imidazolin-2-yl-, dihydrochloride
 :
 :577.5000 g/mol
Storage Temperature:Ambient
CAS Number:6823-69-4
Formula:C₃₀H₃₀Cl₂N₆O₂
MDL Number:MFCD06411564