Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:71-44-3
:
Formula:C₁₀H₂₆N₄
MDL Number:MFCD00008215
:202.35 g/mol
Boiling Pt:150 °C/5mmHg
Melting Pt:28…30 °C
Density:0.937 g/cm³ (25 °C)
Flash Pt:110 °C
Storage Temperature:Refrigerator
EINECS:200-754-2
UN:3259
ADR:8,II
:GHS05
Frequently Bought Together