
N-(tert-Butoxycarbonyl)-L-phenylalanine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13734-34-4
:
Formula:C₁₄H₁₉NO₄
MDL Number:MFCD00002663
:265.31 g/mol
Melting Pt:85…87 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together