Order Entry
Italy
Orders LinkContactUsLinkComponent
L(+)-Histidine ≥99%

L(+)-Histidine ≥99%

Irritant
Supplier:   Apollo Scientific
L(+)-Histidine ≥99%
Supplier:  Apollo Scientific
Certificates
L(+)-Histidine ≥99%

2 Options Available Below

Product Details & Documents

USP, Pyrogen free

Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Melting Pt:282 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-745-3

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.