

Tetracycline hydrochloride 10 mg/ml
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64-75-5
:
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142
:480.9 g/mol
:GHS07
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together