EDTA disodium salt dihydrate ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
:372.24 g/mol
Melting Pt:248 °C (dec.)
EINECS:205-358-3
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together