
(+)-1-Deoxynojirimycin 99% min
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19130-96-2
:
Formula:C₆H₁₃NO₄
MDL Number:MFCD00063474
:163.17 g/mol
Melting Pt:195…202 °C
EINECS:606-239-2
:GHS07
Storage Temperature:Freezer
Frequently Bought Together