
Sulpho-LC-SPDP (Sulphosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)
Catalog # APOSBICL209-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:527.567 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:169751-10-4
Formula:C₁₈H₂₂N₃NaO₈S₃
MDL Number:MFCD00083162