Chromomycin A3
Supplier: Apollo Scientific
Jump to:
About this item
Antibiotic that can serve as a fluorescent DNA stain.
- CAS Number:7059-24-7
- Formula:C₅₇H₈₂O₂₆
- MDL Number:MFCD00043151
- Molecular Weight:1183.30 g/mol
- Melting Pt:182 °C
- Storage Temperature:Freezer
- EINECS:230-348-0
- UN:3462
- ADR:6.1,I
5 Options Available Below
Product Details & Documents
Antibiotic that can serve as a fluorescent DNA stain.
- CAS Number:7059-24-7
- Formula:C₅₇H₈₂O₂₆
- MDL Number:MFCD00043151
- Molecular Weight:1183.30 g/mol
- Melting Pt:182 °C
- Storage Temperature:Freezer
- EINECS:230-348-0
- UN:3462
- ADR:6.1,I
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



