N-Fmoc-N-succinyl-4,7,10-trioxa-1,13-tridecanediamine ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:172089-14-4
:
Formula:C₂₉H₃₈N₂O₈
MDL Number:MFCD05663769
:542.6 g/mol
Storage Temperature:Freezer
Frequently Bought Together