N-1-Fmoc-1,5-diaminopentane · HCl ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:118119-32-7
:
Formula:C₂₀H₂₄N₂O₂·HCl
MDL Number:MFCD00832511
:360.88 g/mol
Storage Temperature:Ambient
Frequently Bought Together
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol, SupraSolv® for GC-MS
Supplier: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals