Order Entry
Italy
ContactUsLinkComponent
Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate
Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate
Catalog # APOSBIB6068-25G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate
Catalog # APOSBIB6068-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 5-Hydroxy-2-methylindole-3-carboxylic acid ethyl ester, Ethyl 5-hydroxy-2-methyl-1H-indole-3-carboxylate
 :
 :219.24 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:7598-91-6
Formula:C₁₂H₁₃NO₃
MDL Number:MFCD00044575
Melting Pt:205…208 °C