Order Entry
Italy
ContactUsLinkComponent
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98% (oxidised form)
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98% (oxidised form)
Catalog # APOSBIB3013-1G
Supplier:  Apollo Scientific
CAS Number:  
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98% (oxidised form)
Catalog # APOSBIB3013-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:175…178 °C
Storage Temperature:Freezer
EINECS:214-664-6
CAS Number:698999-85-8
Formula:C₂₁H₂₇N₇NaO₁₇P₃·xH₂O
MDL Number:MFCD00150480