
(S)-(+)-Abscisic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21293-29-8
:
Formula:C₁₅H₂₀O₄
MDL Number:MFCD00066545
:264.32 g/mol
Melting Pt:186…188 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together