NANOCOLOR® Photometric Tube Test Kit for Aluminium
MANA985098
:
- Chemical method:Eriochrome Cyanine R
- Pk:1 KIT
- Description:NANOCOLOR® Aluminium 07, test no. 0-98
- Dimensions:160×110×119 mm (including packaging)
- Model:MANA985098
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:372,48 g (including packaging)
- For unit:UV/VIS II;VIS II;500 D;PF-12Plus;Advance;MT Easy VIS;MT Easy UV
- Tests per kit:19
- Range:0,02 - 0,70 mg/L (Al³⁺)
A pre-dosed NANOCOLOR tube test for photometric determination of aluminium in water.
Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well."
Sea water analysis: Yes
Hazardous material: No
Remark: Measuring range on NANOCOLOR VIS II
: Rugged box with 19 test tubes.