
2-Hydroxy-4-(trifluoromethoxy)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1071156-25-6
:
Formula:C₈H₅F₃O₃
MDL Number:MFCD18397426
:206.119 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together