Order Entry
Italy
ContactUsLinkComponent
4-(Trifluoromethoxy)benzene-1,3-diol 95%
4-(Trifluoromethoxy)benzene-1,3-diol 95%
Catalog # APOSPC910300-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Trifluoromethoxy)benzene-1,3-diol 95%
Catalog # APOSPC910300-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.108 g/mol
Storage Temperature:Ambient
CAS Number:1395038-04-6
Formula:C₇H₅F₃O₃
MDL Number:MFCD23111252