1-Chloro-4,5-difluoro-2-methoxybenzene 95%
Catalog # APOSPC910121-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.564 g/mol
Storage Temperature:Ambient
CAS Number:1261598-22-4
Formula:C₇H₅ClF₂O
MDL Number:MFCD18415542