5,5-Difluoro-2-azaspiro[3.3]heptane trifluoroacetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1523617-85-7
:
Formula:C₆H₉F₂N·CF₃CO₂H
MDL Number:MFCD26959152
:247.163 g/mol
Storage Temperature:Ambient
Frequently Bought Together