Order Entry
Italy
ContactUsLinkComponent
5-Methoxy-2-(trifluoromethyl)benzaldehyde 95%
5-Methoxy-2-(trifluoromethyl)benzaldehyde 95%
Catalog # APOSPC905233-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Methoxy-2-(trifluoromethyl)benzaldehyde 95%
Catalog # APOSPC905233-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :204.146 g/mol
Storage Temperature:Ambient
CAS Number:944905-42-4
Formula:C₉H₇F₃O₂
MDL Number:MFCD10696315