Order Entry
Italy
ContactUsLinkComponent
Methyl 5-cyano-2-fluorobenzoate 95%
Methyl 5-cyano-2-fluorobenzoate 95%
Catalog # APOSPC901594-10G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5-cyano-2-fluorobenzoate 95%
Catalog # APOSPC901594-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :179.148 g/mol
Storage Temperature:Ambient
CAS Number:337362-21-7
Formula:C₉H₆FNO₂
MDL Number:MFCD11042790