Methyl 2-nitro-4-(trifluoromethyl)phenylacetate
Catalog # APOSPC7975-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:263.17 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:13544-07-5
Formula:C₁₀H₈F₃NO₄
MDL Number:MFCD09800854