0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
2-(2-Bromo-4-fluorophenoxy)propane
2-(2-Bromo-4-fluorophenoxy)propane 98%
2-(2-Bromo-4-fluorophenoxy)propane 98%
2-(2-Bromo-4-fluorophenoxy)propane 98%
APOSPC7408-1G
:
Apollo Scientific
:
2-(2-Bromo-4-fluorophenoxy)propane 98%
APOSPC7408-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
233.078 g/mol
:
243 °C (760 mm Hg)
:
GHS07
:
Ambient
:
202865-79-0
:
C₉H₁₀BrFO
:
MFCD00070761