
1,1,2-Trifluoro-2-chloroethyl 2,2,2-trifluoroethyl ether 97%
Catalog # APOSPC7292D-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.509 g/mol
Boiling Pt:81 °C
Density:1.532 g/cm³ (25 °C)
:GHS07
Storage Temperature:Refrigerator
CAS Number:25364-98-1
Formula:C₄H₃ClF₆O
MDL Number:MFCD00236120