Order Entry
Italy
ContactUsLinkComponent
1,5-Difluoro-2,4-dimethoxybenzene 97%
Irritant
1,5-Difluoro-2,4-dimethoxybenzene 97%
Catalog # APOSPC6689-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,5-Difluoro-2,4-dimethoxybenzene 97%
Catalog # APOSPC6689-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.145 g/mol
Boiling Pt:55 °C (50 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:79069-70-8
Formula:C₈H₈F₂O₂
MDL Number:MFCD13193047