Order Entry
Italy
ContactUsLinkComponent
1H,1H,2H,2H-Perfluorododecyl acrylate 97%
Irritant
Environment
1H,1H,2H,2H-Perfluorododecyl acrylate 97%
Catalog # APOSPC5969O-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
1H,1H,2H,2H-Perfluorododecyl acrylate 97%
Catalog # APOSPC5969O-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :618.181 g/mol
Boiling Pt:122 °C (4 mm Hg)
Melting Pt:48…53 °C
Density:1.32 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:241-732-2
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:17741-60-5
Formula:C₁₅H₇F₂₁O₂
MDL Number:MFCD00236108