Order Entry
Italy
ContactUsLinkComponent
3H-Perfluoro(2-methylbut-2-ene) 97%
3H-Perfluoro(2-methylbut-2-ene) 97%
Catalog # APOSPC5739-5G
Supplier:  Apollo Scientific
CAS Number:  
3H-Perfluoro(2-methylbut-2-ene) 97%
Catalog # APOSPC5739-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Cylinder
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :232.047 g/mol
Storage Temperature:Ambient
CAS Number:22692-37-1
Formula:C₅HF₉
MDL Number:MFCD10565607