0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
3-[3-(Trifluoromethyl)phenyl]propanal
3-[3-(Trifluoromethyl)phenyl]propanal
3-[3-(Trifluoromethyl)phenyl]propanal
APOSPC57040-1G
:
Apollo Scientific
:
3-[3-(Trifluoromethyl)phenyl]propanal
APOSPC57040-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
202.173 g/mol
:
GHS07
:
Ambient
:
21172-41-8
:
C₁₀H₉F₃O
:
MFCD07772918