
Ethyl 6,7-difluoro-8-methoxy-4-oxo-1H-quinoline-3-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:950726-72-4
:
Formula:C₁₃H₁₁F₂NO₄
MDL Number:MFCD31742295
:283.228 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together