Order Entry
Italy
ContactUsLinkComponent
4,4,5,5-Tetrachloro-2,2-difluoro-1,3-dioxolane
Flammable
Toxic
4,4,5,5-Tetrachloro-2,2-difluoro-1,3-dioxolane
Catalog # APOSPC56232-1G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Toxic
4,4,5,5-Tetrachloro-2,2-difluoro-1,3-dioxolane
Catalog # APOSPC56232-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.84 g/mol
Storage Temperature:Ambient
UN:1992
ADR:3+6.1,III
 :GHS02
 :GHS06
CAS Number:87075-01-2
Formula:C₃Cl₄F₂O₂
MDL Number:MFCD28054322