Order Entry
Italy
ContactUsLinkComponent
2-[(4-Bromophenoxy)methyl]-1,1,3,3,3-pentafluoro-1-propene
Irritant
2-[(4-Bromophenoxy)methyl]-1,1,3,3,3-pentafluoro-1-propene
Catalog # APOSPC56228-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-[(4-Bromophenoxy)methyl]-1,1,3,3,3-pentafluoro-1-propene
Catalog # APOSPC56228-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :317.05 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1980085-16-2
Formula:C₁₀H₆BrF₅O
MDL Number:MFCD28334477