3-(Trifluoromethylsulphonyl)phenylacetic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1301739-14-9
:
Formula:C₉H₇F₃O₄S
MDL Number:MFCD25460318
:268.21 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together