2,2-Difluoro-5-(hydroxymethyl)-1,3-benzodioxole
Catalog # APOSPC5472-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.128 g/mol
Storage Temperature:Ambient
CAS Number:72768-97-9
Formula:C₈H₆F₂O₃
MDL Number:MFCD06202699