Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-fluoro-4-formamidobenzoate
Methyl 3-fluoro-4-formamidobenzoate ≥98%
Methyl 3-fluoro-4-formamidobenzoate ≥98%
APOSPC540074-1G
:
Apollo Scientific
:
Methyl 3-fluoro-4-formamidobenzoate ≥98%
APOSPC540074-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
197.163 g/mol
:
Ambient
:
1314936-23-6
:
C₉H₈FNO₃
:
MFCD18642899