
3-(2,2,2-Trifluoroethoxy)benzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1365941-75-8
:
Formula:C₈H₆ClF₃O₃S
MDL Number:MFCD21601592
:274.645 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together