
1-(4-Fluorophenyl)-N3,N3-dimethylpropane-1,3-diamine dihydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2149602-34-4
:
Formula:C₁₁H₁₇FN₂·2ClH
MDL Number:MFCD30492327
:269.186 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together