


Difluoromethyl trifluoromethanesulphonate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1885-46-7
:
Formula:C₂HF₅O₃S
MDL Number:MFCD28054469
:200.085 g/mol
Boiling Pt:49 °C
Density:1.584 g/cm³ (25 °C)
Flash Pt:57 °C
UN:2920
ADR:8+3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together