Order Entry
Italy
ContactUsLinkComponent
(S)-1-(6-(Trifluoromethyl)pyridin-3-yl)ethanamine
Irritant
(S)-1-(6-(Trifluoromethyl)pyridin-3-yl)ethanamine
Catalog # APOSPC53142-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-1-(6-(Trifluoromethyl)pyridin-3-yl)ethanamine
Catalog # APOSPC53142-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :190.166 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1071435-62-5
Formula:C₈H₉F₃N₂
MDL Number:MFCD09256747