(S)-1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride 95+%
Catalog # APOSPC53093-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:241.638 g/mol
Storage Temperature:Ambient
CAS Number:1391540-47-8
Formula:C₉H₁₀F₃NO·ClH
MDL Number:MFCD12757222