Order Entry
Italy
ContactUsLinkComponent
 
Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 95%
  APOSPC53080-1G
 :  
Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 95%
  APOSPC53080-1G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :285.112 g/mol
 :Ambient
 :926280-84-4
 :C₁₀H₁₂BF₃NO₂·K
 :MFCD10698612