
2-Fluoro-5-iodo-4-methylphenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1935452-03-1
:
Formula:C₈H₅F₄IO₃S
MDL Number:MFCD28963744
ADR:8,III
:384.087 g/mol
UN:3265
:GHS05
Storage Temperature:Ambient
Frequently Bought Together