
3-Bromo-2-chloro-6-fluorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1114809-13-0
:
Formula:C₇H₃BrClFO₂
MDL Number:MFCD11856020
Melting Pt:134…136 °C
:253.453 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together