
2-Chloro-5-fluoro-4-(2-hydroxyethylamino)pyrimidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16255-90-6
:
Formula:C₆H₇ClFN₃O
MDL Number:MFCD02629720
Melting Pt:148…151 °C
Flash Pt:>110 °C
UN:2811
ADR:6.1,III
:191.591 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together